Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFBKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637002.3 | XP_005637059.1 | 3 - 4 | 776 | 86 | |
XM_005637004.3 | XP_005637061.1 | 2 - 3 | 423 | 86 | ||
XM_022410992.1 | XP_022266700.1 | 5 - 6 | 1494 | 86 | ||
XM_022410993.1 | XP_022266701.1 | 4 - 5 | 954 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |