Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080899.1 | NP_001074368.1 | 4 - 5 | 357 | 111 | |
GenBank mRNA | EF191198.1 | - | 4 - 5 | 357 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |