Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632459.3 | XP_005632516.1 | 4 - 5 | 852 | 85 | |
XM_005632460.3 | XP_005632517.1 | 5 - 6 | 950 | 85 | ||
XM_014121936.2 | XP_013977411.1 | 4 - 5 | 892 | 85 | ||
XM_022406646.1 | XP_022262354.1 | 5 - 6 | 932 | 85 | ||
XM_022406647.1 | XP_022262355.1 | 4 - 5 | 855 | 85 | ||
XM_022406648.1 | XP_022262356.1 | 5 - 6 | 1238 | 85 | ||
XM_022406649.1 | XP_022262357.1 | 4 - 5 | 942 | 85 | ||
XM_541854.6 | XP_541854.1 | 4 - 5 | 859 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |