Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: MEPCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419643.1 | XP_022275351.1 | 1310 | 107 | ||
XM_546957.6 | XP_546957.2 | 1731 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |