Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425831.1 | XP_022281539.1 | 1 | 557 | 110 | |
XM_532065.6 | XP_532065.1 | 2 | 642 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |