Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_843970.5 | XP_849063.2 | 4 | 682 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |