Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284494.1 | NP_001271423.1 | 4 | 513 | 195 | |
GenBank mRNA | JQ305104.1 | - | 4 | 513 | 195 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Both primers and probe map within a single exon |