Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622671.3 | XP_005622728.2 | 4 | 520 | 97 | |
XM_022421225.1 | XP_022276933.1 | 4 | 512 | 97 | ||
XM_022421226.1 | XP_022276934.1 | 4 | 512 | 97 | ||
XM_537241.5 | XP_537241.3 | 4 | 512 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |