Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119722.2 | XP_013975197.1 | 2 | 703 | 152 | |
XM_022403249.1 | XP_022258957.1 | 2 | 823 | 152 | ||
XM_022403250.1 | XP_022258958.1 | 2 | 660 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |