Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003254.2 | NP_001003254.1 | 4 - 5 | 496 | 76 | |
GenBank mRNA | Z49944.1 | - | 496 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |