Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252291.1 | NP_001239220.1 | 1 | 174 | 98 | |
GenBank mRNA | DN398351.1 | - | 1 | 174 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |