Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244037.1 | NP_001230966.1 | 2 - 3 | 187 | 80 | |
GenBank mRNA | JI886339.1 | - | 1 - 2 | 80 | ||
U55387.2 | - | 2 - 3 | 187 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |