Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244035.1 | NP_001230964.1 | 1 | 814 | 89 | |
XM_007646456.1 | XP_007644646.1 | 3 | 1120 | 89 | ||
XM_007646457.1 | XP_007644647.1 | 2 | 990 | 89 | ||
GenBank mRNA | D30618.1 | - | 1 | 814 | 89 | |
Z17220.1 | - | 1 | 814 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |