Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509765.2 | XP_003509813.1 | 4 - 5 | 1159 | 57 | |
XM_007650419.1 | XP_007648609.1 | 4 - 5 | 1064 | 57 | ||
XM_007650420.1 | XP_007648610.1 | 4 - 5 | 1155 | 57 | ||
XM_007650421.1 | XP_007648611.1 | 4 - 5 | 1203 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |