Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508785.2 | XP_003508833.1 | 1 | 417 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Both primers and probe map within a single exon |