Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648890.1 | XP_007647080.1 | 3 - 4 | 442 | 74 | |
GenBank mRNA | JI885869.1 | - | 3 - 4 | 424 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |