Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Llph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497127.2 | XP_003497175.1 | 2 - 3 | 296 | 93 | |
GenBank mRNA | JI889203.1 | - | 2 - 3 | 317 | 93 | |
JP057428.1 | - | 2 - 3 | 317 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |