Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510791.2 | XP_003510839.2 | 1 - 2 | 456 | 74 | |
GenBank mRNA | JI864864.1 | - | 1 - 2 | 196 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |