Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500324.2 | XP_003500372.2 | 2 - 3 | 160 | 62 | |
XM_007641054.1 | XP_007639244.1 | 2 - 3 | 492 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |