Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513659.2 | XP_003513707.1 | 12 - 13 | 2612 | 89 | |
XM_007653918.1 | XP_007652108.1 | 12 - 13 | 2542 | 89 | ||
XM_007653919.1 | XP_007652109.1 | 12 - 13 | 2709 | 89 | ||
XM_007653920.1 | XP_007652110.1 | 9 - 10 | 2030 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |