Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505073.2 | XP_003505121.2 | 3 - 4 | 254 | 85 | |
GenBank mRNA | JI894944.1 | - | 3 - 4 | 239 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |