Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514274.2 | XP_003514322.2 | 4 - 5 | 604 | 77 | |
GenBank mRNA | JP055849.1 | - | 4 - 5 | 604 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |