Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aste1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497633.2 | XP_003497681.1 | 4 - 5 | 2007 | 69 | |
XM_007615046.1 | XP_007613236.1 | 2 - 3 | 243 | 69 | ||
GenBank mRNA | JI886198.1 | - | 1 - 2 | 194 | 69 | |
JP043295.1 | - | 3 - 4 | 1154 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |