Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679509.3 | XP_005679566.1 | 2 - 3 | 532 | 68 | |
XM_005679510.3 | XP_005679567.1 | 2 - 3 | 537 | 68 | ||
GenBank mRNA | JO582023.1 | - | 3 - 4 | 1153 | 68 | |
JO633022.2 | - | 3 - 4 | 820 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |