Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692713.3 | XP_005692770.1 | 3 - 4 | 489 | 75 | |
XM_005692714.3 | XP_005692771.1 | 3 - 4 | 423 | 75 | ||
XM_005692715.2 | XP_005692772.1 | 3 - 4 | 364 | 75 | ||
XM_013971503.2 | XP_013826957.1 | 3 - 4 | 431 | 75 | ||
XM_013971504.2 | XP_013826958.1 | 3 - 4 | 413 | 75 | ||
XM_013971505.2 | XP_013826959.1 | 3 - 4 | 489 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |