Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062760.1 | XP_017918249.1 | 4 - 5 | 501 | 64 | |
GenBank mRNA | JO622411.2 | - | 4 - 5 | 525 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |