Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051099.1 | XP_017906588.1 | 4 - 5 | 525 | 76 | |
GenBank mRNA | JO576441.1 | - | 1 - 2 | 836 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |