Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687038.2 | XP_005687095.1 | 1 - 2 | 186 | 69 | |
GenBank mRNA | JO642951.2 | - | 1 - 2 | 170 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |