Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287041.1 | NP_001273970.1 | 2 - 3 | 1295 | 54 | |
NM_001287042.1 | NP_001273971.1 | 2 - 3 | 330 | 54 | ||
GenBank mRNA | DQ645739.1 | - | 2 - 3 | 194 | 54 | |
GQ246165.1 | - | 1 - 2 | 160 | 54 | ||
JF896277.1 | - | 1 - 2 | 157 | 54 | ||
JF896278.1 | - | 1 - 2 | 157 | 54 | ||
JO343911.2 | - | 2 - 3 | 1295 | 54 | ||
JO421020.2 | - | 3 - 4 | 509 | 54 | ||
JO421021.2 | - | 4 - 5 | 674 | 54 | ||
KJ413092.1 | - | 1 - 2 | 157 | 54 | ||
KT445943.1 | - | 2 - 3 | 187 | 54 | ||
KT445944.1 | - | 2 - 3 | 187 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |