Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AOPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052467.1 | XP_017907956.1 | 5 - 6 | 1575 | 79 | |
XM_018052468.1 | XP_017907957.1 | 5 - 6 | 1575 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |