Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059511.1 | XP_017915000.1 | 2 - 3 | 749 | 110 | |
GenBank mRNA | JO369533.2 | - | 3 - 4 | 620 | 110 | |
JO577012.1 | - | 4 - 5 | 1043 | 110 | ||
JO577013.1 | - | 4 - 5 | 1043 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |