Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677108.3 | XP_005677165.3 | 2 - 3 | 534 | 108 | |
GenBank mRNA | JO511845.2 | - | 2 - 3 | 537 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |