Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677804.3 | XP_005677861.2 | 2 - 3 | 165 | 79 | |
XM_013962499.2 | XP_013817953.2 | 2 - 3 | 165 | 79 | ||
GenBank mRNA | KA328838.1 | - | 2 - 3 | 200 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |