Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057633.1 | XP_017913122.1 | 3 - 4 | 833 | 85 | |
XM_018057635.1 | XP_017913124.1 | 4 - 5 | 1068 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |