Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056478.1 | XP_017911967.1 | 4 - 5 | 1542 | 71 | |
XM_018056479.1 | XP_017911968.1 | 4 - 5 | 775 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |