Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055323.1 | XP_017910812.1 | 3 - 4 | 538 | 58 | |
XM_018055325.1 | XP_017910814.1 | 1 - 2 | 243 | 58 | ||
GenBank mRNA | JO349095.2 | - | 3 - 4 | 579 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |