Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692684.3 | XP_005692741.2 | 5 - 6 | 1322 | 59 | |
XM_018062726.1 | XP_017918215.1 | 5 - 6 | 1327 | 59 | ||
XM_018062727.1 | XP_017918216.1 | 4 - 5 | 1135 | 59 | ||
GenBank mRNA | JO633226.2 | - | 5 - 6 | 1263 | 59 | |
JO633228.2 | - | 5 - 6 | 1263 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |