Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690983.3 | XP_005691040.1 | 8 - 9 | 895 | 117 | |
XM_005690986.2 | XP_005691043.1 | 7 - 8 | 799 | 117 | ||
XM_005690987.2 | XP_005691044.1 | 7 - 8 | 773 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |