Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050406.1 | XP_017905895.1 | 3 - 4 | 273 | 77 | |
XM_018050407.1 | XP_017905896.1 | 3 - 4 | 240 | 77 | ||
GenBank mRNA | JO632047.2 | - | 3 - 4 | 266 | 77 | |
KA332497.1 | - | 4 - 5 | 1914 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |