Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTREX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694702.3 | XP_005694759.2 | 2 - 3 | 460 | 66 | |
GenBank mRNA | JO424332.1 | - | 30 - 31 | 4639 | 66 | |
JO424740.1 | - | 30 - 31 | 4642 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() |