Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699784.3 | XP_005699841.1 | 3 - 4 | 543 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() |