Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064021.1 | XP_017919510.1 | 1 - 2 | 250 | 78 | |
GenBank mRNA | JO431978.2 | - | 5 - 6 | 1519 | 78 | |
JT677007.1 | - | 6 - 7 | 1650 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |