Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H16orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974638.1 | XP_013830092.1 | 1 | 516 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |