Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677243.3 | XP_005677300.2 | 2 - 3 | 137 | 99 | |
GenBank mRNA | JO613738.2 | - | 8 - 9 | 1352 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |