Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041454.1 | XP_017896943.1 | 3 - 4 | 790 | 55 | |
GenBank mRNA | KA348697.1 | - | 2 - 3 | 127 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |