Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050508.1 | XP_017905997.1 | 5 - 6 | 1648 | 65 | |
GenBank mRNA | JO632303.2 | - | 3 - 4 | 1746 | 65 | |
JT639651.1 | - | 5 - 6 | 1107 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |