Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: spict | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273492.1 | NP_001260421.1 | 1 - 2 | 371 | 57 | |
NM_135742.4 | NP_609586.1 | 1 - 2 | 371 | 57 | ||
GenBank mRNA | AY089521.1 | - | 1 - 2 | 351 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |