Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136429.4 | NP_610273.1 | 5 - 6 | 1364 | 88 | |
NM_165518.3 | NP_724547.1 | 5 - 6 | 1966 | 88 | ||
NM_165519.3 | NP_724548.1 | 4 - 5 | 2043 | 88 | ||
NM_165521.3 | NP_724550.1 | 5 - 6 | 912 | 88 | ||
GenBank mRNA | BT001503.1 | - | 4 - 5 | 2019 | 88 | |
BT001556.1 | - | 5 - 6 | 1353 | 88 | ||
BT003263.1 | - | 5 - 6 | 855 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |