Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057750.4 | NP_477098.1 | 4 - 5 | 657 | 119 | |
NM_134308.3 | NP_599135.1 | 4 - 5 | 657 | 119 | ||
NM_134309.2 | NP_599136.1 | 4 - 5 | 528 | 119 | ||
NM_166139.3 | NP_725524.1 | 3 - 4 | 539 | 119 | ||
NM_206127.2 | NP_995849.1 | 4 - 5 | 606 | 119 | ||
NM_206128.2 | NP_995850.1 | 4 - 5 | 600 | 119 | ||
GenBank mRNA | AF177872.1 | - | 4 - 5 | 610 | 119 | |
AF177873.1 | - | 4 - 5 | 565 | 119 | ||
AF177874.2 | - | 4 - 5 | 427 | 119 | ||
AY119536.1 | - | 3 - 4 | 523 | 119 | ||
BT010085.1 | - | 4 - 5 | 599 | 119 | ||
L38311.1 | - | 427 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |