Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lis-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259432.2 | NP_001246361.1 | 3 - 4 | 466 | 63 | |
NM_057812.5 | NP_477160.1 | 3 - 4 | 541 | 63 | ||
NM_176190.2 | NP_788370.1 | 3 - 4 | 454 | 63 | ||
NM_206130.2 | NP_995852.1 | 3 - 4 | 971 | 63 | ||
GenBank mRNA | AF117606.1 | - | 3 - 4 | 545 | 63 | |
AF152419.1 | - | 3 - 4 | 530 | 63 | ||
BT021404.1 | - | 3 - 4 | 536 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |